Orders LinkContactUsLinkComponent
Tank pump sampler set
Supplier:   Sampling Systems
Tank pump sampler set
Supplier:  Sampling Systems
Certificates
Tank pump sampler set
Tank pump sampler set
2 Options Available Below
Product Details & Documents
Perfect for sampling low viscosity liquids.
  • Quick and convenient sampling of liquids
  • Sample from depths of approximately 4 m
Sampling low viscosity liquids such as waters and oils is quick and easy with the Tank Pump sampler set. The only product contact parts are the dip tube and sample collection bottle. The hand pump operates with tube outer diameters between 5 and 8 mm.
 : Each set includes a hand pump, 12 PE bottles (150 ml set) or 5 PE bottles (250 ml set), a 13 m PE tube, a tube cutter, pen, and carrying case.
Product Family Options
Product Information
  • Descrizione
    Capacità
    Availability
    Price
  • Tank pump sampler set
    150 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 319.00
    SFr. 319.00
    1 * 1 ST
       SFr. 319.00
  • Tank pump sampler set
    250 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 319.00
    SFr. 319.00
    1 * 1 ST
       SFr. 319.00