Specifications
- Larghezza:1219 mm
- Colore:trasparente
- Descrizione:Removable repositionable double coated tape, 9425HT
- Lunghezza:55 m
- Conf:1 Roll
Specifications
About this item
This 152 µm double coated tape with high tack acrylic adhesive 420 on one side and medium tack acrylic adhesive 1050 on the other side, is supplied on a 58 lbs. polycoated kraft paper liner with a PET carrier.
- 1,4 mil PET carrier coated with 2,2 mil 420 adhesive on face side and 2,2 mil 1050 adhesive on back side
- Suitable for high temperature applications
- Position and reposition lightweight materials
- Works on a variety of substrates or materials for temporary mounting of lightweight materials
Temporarily fix or mount lightweight materials or create multiple opening and closing applications with 3M™ Removable Repositionable Double Coated Tape 9425HT. Reposition a variety of substrates or materials such as paper, plastics, films, or foams. Laminate and die cut with ease using the UPVC film carrier, which gives differential stability. The PET film carrier provides differential stability which offers improved handling for laminating and die cutting.
This double coated adhesive tape has high tack acrylic adhesive on one side and medium tack acrylic adhesive on the other side, on a polycoated kraft paper liner with a UPVC carrier.