Conductive tips, MBP
Tecan style art tips, 1000 µl, +, Nero
Codice catalogo MLBP904-013
Nome del fornitore: Thermo Fisher Scientific
Conductive tips, MBP
Codice catalogo MLBP904-013
Nome del fornitore: Thermo Fisher Scientific
Vai a
Product Details & Documents
Filtered, conductive tip style, liquid sensing, Econopacks.
- For use with: Tecan™ Freedom EVO™ LiHa Head, Tecan™ Genesis™
- Ideal for automated liquid handling systems
- Non sterile, polypropylene
Perkin-Elmer is a registered trademark of Perkin-Elmer Corporation. ART is a registered trademark of MBP and is protected by U.S. Patent # 5,156,811. EconoPak is a trademark of Molecular BioProducts. Tecan is a registered trademark of Tecan AG. Cavro is a registered trademark of Scientific Instruments, Inc. MultiPROBE and Packard are registered trademarks of Packard Instrument Company, Inc. Thermo Orion is a registered trademark of Thermo Orion Inc.
Specifiche
- Volume:1000 µl
- Stile punta:Tecan style art tips
- Colore puntale:Nero
- Filtro:+
- Cat. No.:MLBP904-013
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)