Powered air respirator air duct sealing rings, Versaflo™
Catalog # MMMAS-956
Supplier: 3M
Specifications
- Descrizione:Air duct sealing rings
- Conf:5
Specifications
About this item
The Versaflo™ Air Duct Sealing Ring S-956 is a spare air duct sealing ring designed to replace worn or damaged parts in Versaflo™ S-Series Premium S-950 Head Suspension.
- Replacement air duct sealing ring
- Approved 3M replacement part
- Extend product life by regularly checking for and replacing worn or damaged parts
This replacement part can extend the service life of head suspensions. The sealing ring is installed on the section where air enters the suspension.