Powered and supplied air sweatbands and comfort pads, Versaflo™
Codice catalogo MMMAM956
Nome del fornitore: 3M
Specifiche
- Descrizione:Sweatbands and comfort pads
- Conf:10
Specifiche
Informazioni su questo articolo
The Versaflo™ Size Reducing Ratchet Comfort Pad M-956 is a ratchet-style comfort pad that fits to the back of a 3M™ Versaflo™ M-Series Headtop to reduce the size of the headtop.
- Size reducing comfort pad fits to rachet of helmet
- Adds comfort and helps fit smaller wearers
- Designed for use with 3M™ Versaflo™ M-Series Headtops
Adjust the fit of 3M™ Versaflo™ M-Series Headtop with a Versaflo™ Size Reducing Ratchet Comfort Pad, M-956. It’s easy to fit comfort pads onto the rachet at the back of a helmet to help fit smaller wearers or provide additional comfort to any user.