Powered and supplied air breathing tubes
Catalog # MMMAS222
Supplier: 3M
Specifications
- Descrizione:Powered and supplied air breathing tubes
- Seq:03
- Conf:1
Specifications
About this item
Breathing Tube S-222 is a replacement breathing tube for use with 3M™ Versaflo™ Supplied Air System S-200+.
- Spare or replacement part
- Protective breathing tube covers available to lengthen service life of breathing tubes
Prolong the service life of your 3M™ Versaflo™ Supplied Air System S-200+ with the Breathing Tube, S-222. This spare breathing tube is specially designed for use with S-200+ systems and is CE approved.