Compartment Boxes
Informazioni su questo articolo
Compartment boxes are suitable for storage, transport, and presentation of small parts. All models are provided with a perfectly transparent cover in order to showcase the contents inside.
- High transparency lid offers optimum visibility of contents
- 100% recyclable virgin material
- Box material: Butadiene styrene copolymer
- Inserts material: Polystyrene
- Transparent cover with secure closure by simple pressure
- Perfect stacking stability
The boxes are equipped with white removable inserts. They are made from high quality materials that are strong and durable. The thickness of the material, the careful design, and the quality of design make them excellent presentation boxes for professionals.
10 varianti
Product Details & Documents
Compartment boxes are suitable for storage, transport, and presentation of small parts. All models are provided with a perfectly transparent cover in order to showcase the contents inside.
- High transparency lid offers optimum visibility of contents
- 100% recyclable virgin material
- Box material: Butadiene styrene copolymer
- Inserts material: Polystyrene
- Transparent cover with secure closure by simple pressure
- Perfect stacking stability
The boxes are equipped with white removable inserts. They are made from high quality materials that are strong and durable. The thickness of the material, the careful design, and the quality of design make them excellent presentation boxes for professionals.
Product Family Options
Informazioni sul prodotto
- L×P×ADisponibilitàPrice
- 216-248715-compartment box, with 45×45 mm compartment sizes15-compartment box, with 45×45 mm compartment sizes285×177×25 mmLog in per disponibilità1 * 10 STSFr. 258.00
- 216-248831-compartment box, with 150×45 / 31×26 mm compartment sizes31-compartment box, with 150×45 / 31×26 mm compartment sizes285×177×25 mmLog in per disponibilità1 * 10 STSFr. 258.00
- 216-248954-compartment box, with 25×20 mm compartment sizes54-compartment box, with 25×20 mm compartment sizes285×177×25 mmLog in per disponibilità1 * 10 STSFr. 258.00
- 216-24805-compartment box, with 151×65 / 60×60 / 194×85 mm compartment sizes5-compartment box, with 151×65 / 60×60 / 194×85 mm compartment sizes285×177×45 mmLog in per disponibilità1 * 8 STSFr. 235.00
- 216-24816-compartment box, with 85×73 mm compartment sizes6-compartment box, with 85×73 mm compartment sizes285×177×45 mmLog in per disponibilità1 * 8 STSFr. 235.00
- 216-248212-compartment box, with 62×47 mm compartment sizes12-compartment box, with 62×47 mm compartment sizes285×177×45 mmLog in per disponibilità1 * 8 STSFr. 235.00
- 216-248315-compartment box, with 47×47 mm compartment sizes15-compartment box, with 47×47 mm compartment sizes285×177×45 mmLog in per disponibilità1 * 8 STSFr. 235.00
- 216-248428-compartment box, with 35×35 mm compartment sizes28-compartment box, with 35×35 mm compartment sizes285×177×45 mmLog in per disponibilità1 * 8 STSFr. 235.00
- 216-248531-compartment box, with 31×27 / 45×150 mm compartment sizes31-compartment box, with 31×27 / 45×150 mm compartment sizes285×177×45 mmLog in per disponibilità1 * 8 STSFr. 235.00
- 216-248654-compartment box, with 24×22 mm compartment sizes54-compartment box, with 24×22 mm compartment sizes285×177×45 mmLog in per disponibilità1 * 8 STSFr. 235.00