ContactUsLinkComponent
Counting chamber for corning cell counter, 0,1 mm, Corning®
Counting chamber for corning cell counter, 0,1 mm, Corning®
Catalog # CORN480200
Supplier:  Corning
undefined
Counting chamber for corning cell counter, 0,1 mm, Corning®
Catalog # CORN480200
Supplier:  Corning
Specifications
  • Profondità della camera:
    0,1 mm
  • Descrizione:
    Counting chamber for Corning cell counter, 10 µl capacity
  • Conf:
    1
Specifications
About this item
The Corning counting chamber is a reusable glass cell counting slide for the Corning cell counter.
This gridless slide has a 0,1 mm depth same as a Neubauer Hemocytometer and comes standard with two high quality cover slips. These slides have a notch for easy loading of cell suspension.