A true workhorse centrifuge with versatile rotor options.
- Tube adapter options accommodate tube sizes from 0,5 to 50 ml
- Digital keypad interface is glove-friendly
- Quiet running centrifuge provides a pleasant work setting
This may be the only centrifuge you need—standard four-place swinging buckets accommodate 50 ml tubes and additional rotor and adapter options accommodate almost any tube size for your runs. Two-line digital display shows status, time, rpm and RCF(g), and the 60-minute timer signals when run is complete. Previous settings can be easily repeated for quick testing. Stainless steel interior chamber is easily accessible for cleaning and will withstand years of use.
- Environmentally Preferable:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.