Masterflex® Micro Peristaltic Pumps, Avantor®
Nome del fornitore: Avantor Fluid Handling
Masterflex® Micro Peristaltic Pumps, Avantor®
Nome del fornitore: Avantor Fluid Handling
Certificates
Informazioni su questo articolo
Operate by computer or on your iOS or Android™ device.
- Easily achieve flow ranges down to 0.2 µL/min with a 1.26% reproducibility
- 12-roller head minimizes the pulsation
- Select pump based upon number of channels needed—from one to eight
- Wide choice of peristaltic tube sets (0.13 mm i.d. to 3.17 mm i.d.)
Control these micro pumps through your computer using an available USB port and Pump Network Control™ software. Software† can be used remotely on the same Wi-Fi network using an Apple® iOS or Android device. The Pump Network Control software controls dire
4 varianti
Product Details & Documents
Operate by computer or on your iOS or Android™ device.
- Easily achieve flow ranges down to 0.2 µL/min with a 1.26% reproducibility
- 12-roller head minimizes the pulsation
- Select pump based upon number of channels needed—from one to eight
- Wide choice of peristaltic tube sets (0.13 mm i.d. to 3.17 mm i.d.)
Control these micro pumps through your computer using an available USB port and Pump Network Control™ software. Software† can be used remotely on the same Wi-Fi network using an Apple® iOS or Android device. The Pump Network Control software controls dire
Specifications for Product Family
Specifiche
- Portata di flusso min.:
- 0,2 µl/min
- 0,2 µl/min
- 0,2 µl/min
- 0,2 µl/min
- Approvazione normativa CE:
- Yes
- Yes
- Yes
- Yes
- Self-priming:
- Yes
- Yes
- Yes
- Yes
- Peso di spedizione:
- 2,15 kg (4,75 lbs.)
- 2,15 kg (4,75 lbs.)
- 2,15 kg (4,75 lbs.)
- 2,15 kg (4,75 lbs.)
- Garanzia:
- 1 year
- 1 year
- 1 year
- 1 year
Specifiche
Product Family Options
Informazioni sul prodotto
- Massima velocità del flussoAltezzaLunghezzaLarghezzaPotenzaDisponibilitàPrice
- MFLX74906-04Six Channel precision micro peristaltic pumpSix Channel precision micro peristaltic pump10 ml/min12,5 cm6,4 cm5,8 cm90 - 240 VACLog in per disponibilità1 * 1 STSFr. 3’560.00Specifiche:CE Compliant
More
Specifications
Cat. No.MFLX74906-04Portata di flusso min.0,2 µl/minApprovazione normativa CEYesSelf-primingYesPeso di spedizione2,15 kg (4,75 lbs.)Garanzia1 yearCE Compliant - MFLX74906-00One Channel precision micro peristaltic pumpOne Channel precision micro peristaltic pump10 ml/min12,5 cm6,4 cm5,8 cm90 - 240 VACLog in per disponibilità1 * 1 STSFr. 2’870.00Specifiche:CE Compliant
More
Specifications
Cat. No.MFLX74906-00Portata di flusso min.0,2 µl/minApprovazione normativa CEYesSelf-primingYesPeso di spedizione2,15 kg (4,75 lbs.)Garanzia1 yearCE Compliant - MFLX74906-01Two Channel precision micro peristaltic pumpTwo Channel precision micro peristaltic pump10 ml/min12,5 cm6,4 cm5,8 cm90 - 240 VACLog in per disponibilità1 * 1 STSFr. 3’020.00Specifiche:CE Compliant
More
Specifications
Cat. No.MFLX74906-01Portata di flusso min.0,2 µl/minApprovazione normativa CEYesSelf-primingYesPeso di spedizione2,15 kg (4,75 lbs.)Garanzia1 yearCE Compliant - MFLX74906-03Four Channel precision micro peristaltic pumpFour Channel precision micro peristaltic pump10 ml/min12,5 cm6,4 cm5,8 cm90 - 240 VACLog in per disponibilità1 * 1 STSFr. 3’500.00Specifiche:CE Compliant
More
Specifications
Cat. No.MFLX74906-03Portata di flusso min.0,2 µl/minApprovazione normativa CEYesSelf-primingYesPeso di spedizione2,15 kg (4,75 lbs.)Garanzia1 yearCE Compliant