Bulkhead Fitting Connectors, PTFE
Bulkhead Fitting, PTFE, ⅛" NPT(F) Portto1⅝" OD, Compression to Thread, PTFE, Straight, ⅛"
Codice catalogo MFLX34031-11
Nome del fornitore: Avantor Fluid Handling
Bulkhead Fitting Connectors, PTFE
Codice catalogo MFLX34031-11
Nome del fornitore: Avantor Fluid Handling
Vai a
Product Details & Documents
Suitable for use with flat-wall or most cylindrical, smooth-wall tanks.
- Includes FEP encapsulated silicone O-ring
- Body can be made longer to accommodate thicker tank walls
Constructed of durable PTFE, these Bulkhead Fitting Connectors are ideal for a wide range of applications.
Specifiche
- Tipo di connessione:Compression to Thread
- Flow pattern:Straight
- Thread size NPT(F) (port 2):⅛"
- Descrizione:Bulkhead Fitting, PTFE, ⅛" NPT(F) Portto1⅝" OD
- Fitting material:PTFE
- Cat. No.:MFLX34031-11
- Diametro esterno della paratia:1,625
- Sterilizzazione:Autoclave or ethylene oxide (ETO)
- Garanzia:2 years