Orders LinkContactUsLinkComponent
Masterflex® Variable Area Flowmeter Single Tube Frames, Avantor® | Avantor
Masterflex® Variable Area Flowmeter Single Tube Frames, Avantor®
Nome del fornitore:   Avantor Fluid Handling
Masterflex® Variable Area Flowmeter Single Tube Frames, Avantor®
Nome del fornitore:  Avantor Fluid Handling
Certificates
Masterflex® Variable Area Flowmeter Single Tube Frames, Avantor®
Masterflex® Variable Area Flowmeter Single Tube Frames, Avantor®
7 varianti
Product Details & Documents
Create your own flowmeter systems for complete flexibility.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Use with media for laboratory and industrial applications
  • Required system components for flowmeter systems would be frame, flowtube(s), valve cartridge, and tripod—order each component separately
  • Choose from aluminum or stainless steel wetted parts based on required chemical compatibility
  • Designed with 1/8” NPT(F) connections
Design a flow system to meet your metering, mixing, or monitoring applications. We offer a variety of frames and flowtubes so that you can create the system to meet your exact needs.
Choose a frame depending on the flowtube capacity, flow pattern, and wetted parts that you require.
Choose a frame with a valve port if you plan on adding a valve cartridge to control flow.
Specifications for Product Family
Specifiche
  • Temperatura di esercizio massima:
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • Pressione max.:
  • 200 psi
  • 200 psi
  • 200 psi
  • 200 psi
  • 200 psi
  • 200 psi
  • 200 psi
  • Garanzia:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Specifiche
Product Family Options
Informazioni sul prodotto
  • Valve
    Housing material
    Process connection
    Disponibilità
    Price
  • MFLX03220-00
    Variable-Area Single-Tube Frame, 65-mm Scale; No Valve Port, Aluminum
    Variable-Area Single-Tube Frame, 65-mm Scale; No Valve Port, Aluminum
    No Valve Port
    Aluminium
    0,125" NPT(F)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 66.70
    SFr. 66.70
    1 * 1 ST
       SFr. 66.70
    Specifiche:
    More
    Specifications
    Cat. No.
    MFLX03220-00
    Temperatura di esercizio massima
    121 °C (250 °F)
    Pressione max.
    200 psi
    Garanzia
    1 year
  • MFLX03220-06
    Variable-Area Single-Tube Frame, 150-mm Scale; No Valve Port, Aluminum
    Variable-Area Single-Tube Frame, 150-mm Scale; No Valve Port, Aluminum
    No Valve Port
    Aluminium
    0,125" NPT(F)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 98.80
    SFr. 98.80
    1 * 1 ST
       SFr. 98.80
    Specifiche:
    More
    Specifications
    Cat. No.
    MFLX03220-06
    Temperatura di esercizio massima
    121 °C (250 °F)
    Pressione max.
    200 psi
    Garanzia
    1 year
  • MFLX03220-08
    Variable-Area Single-Tube Frame, 65-mm Scale; Valve Port, Aluminum
    Variable-Area Single-Tube Frame, 65-mm Scale; Valve Port, Aluminum
    Valve Port
    Aluminium
    0,125" NPT(F)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 58.90
    SFr. 58.90
    1 * 1 ST
       SFr. 58.90
    Specifiche:
    More
    Specifications
    Cat. No.
    MFLX03220-08
    Temperatura di esercizio massima
    121 °C (250 °F)
    Pressione max.
    200 psi
    Garanzia
    1 year
  • MFLX03220-10
    Variable-Area Single-Tube Frame, 150-mm Scale; Valve Port, Aluminum
    Variable-Area Single-Tube Frame, 150-mm Scale; Valve Port, Aluminum
    Valve Port
    Aluminium
    0,125" NPT(F)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 98.80
    SFr. 98.80
    1 * 1 ST
       SFr. 98.80
    Specifiche:
    More
    Specifications
    Cat. No.
    MFLX03220-10
    Temperatura di esercizio massima
    121 °C (250 °F)
    Pressione max.
    200 psi
    Garanzia
    1 year
  • MFLX03220-40
    Variable-Area Single-Tube Frame, 65-mm Scale; No Valve Port, Stainless Steel
    Variable-Area Single-Tube Frame, 65-mm Scale; No Valve Port, Stainless Steel
    No Valve Port
    Stainless steel
    0,125" NPT(F)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 184.00
    SFr. 184.00
    1 * 1 ST
       SFr. 184.00
    Specifiche:
    More
    Specifications
    Cat. No.
    MFLX03220-40
    Temperatura di esercizio massima
    121 °C (250 °F)
    Pressione max.
    200 psi
    Garanzia
    1 year
  • MFLX03220-44
    Variable-Area Single-Tube Frame, 65-mm Scale; Valve Port, Stainless Steel
    Variable-Area Single-Tube Frame, 65-mm Scale; Valve Port, Stainless Steel
    Valve Port
    Stainless steel
    0,125" NPT(F)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 184.00
    SFr. 184.00
    1 * 1 ST
       SFr. 184.00
    Specifiche:
    More
    Specifications
    Cat. No.
    MFLX03220-44
    Temperatura di esercizio massima
    121 °C (250 °F)
    Pressione max.
    200 psi
    Garanzia
    1 year
  • MFLX03220-46
    Variable-Area Single-Tube Frame, 150-mm Scale; Valve Port, Stainless Steel
    Variable-Area Single-Tube Frame, 150-mm Scale; Valve Port, Stainless Steel
    Valve Port
    Stainless steel
    0,125" NPT(F)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 217.00
    SFr. 217.00
    1 * 1 ST
       SFr. 217.00
    Specifiche:
    More
    Specifications
    Cat. No.
    MFLX03220-46
    Temperatura di esercizio massima
    121 °C (250 °F)
    Pressione max.
    200 psi
    Garanzia
    1 year