John Guest polypropylene push-to-connect fittings
Nome del fornitore: Avantor Fluid Handling
Certificati
Informazioni su questo articolo
Quick, leakproof connection.
- Polypropylene and EPDM materials meet NSF 51, and NSF 61 requirements for food-grade applications, and is FDA compliant
Push-to-connect fittings offer secure connections and quick disconnection. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semi-rigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing.
Adapters (sold separately) let you permanently modify basic fittings for customized installations.
- Environmentally Preferable:
Specifiche
- Pressione di esercizio massima:
- 150 psi at 70 °F, 60 psi at 140 °F (for water)
- 150 psi at 70 °F, 60 psi at 140 °F (for water)
- 150 psi at 70 °F, 60 psi at 140 °F (for water)
- 150 psi at 70 °F, 60 psi at 140 °F (for water)
- Temperatura massima:
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- Temperatura minima:
- 1 °C (33 °F)
- 1 °C (33 °F)
- 1 °C (33 °F)
- 1 °C (33 °F)
- Seal material:
- EPDM
- EPDM
- EPDM
- EPDM
- Sterilizzazione:
- Autoclave only
- Autoclave only
- Autoclave only
- Autoclave only
- Garanzia:
- 20 years
- 20 years
- 20 years
- 20 years