Masterflex® Adapter Fittings, Luer to Luer, Nylon, Avantor®
Nome del fornitore: Avantor Fluid Handling
Certificati
Informazioni su questo articolo
Easily transition to a luer fitting.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Easily complete your luer fittings systems with these premium connectors. All female ports may be used with either slip or luer lock connections. Plugs and caps are also available.
- Environmentally Preferable:
Specifiche
- Catalog No:
- MFLX45502-84
- MFLX45502-28
- MFLX45502-85
- MFLX45502-22
- MFLX45502-80
- MFLX45505-56
- MFLX45502-56
- MFLX45505-72
- Modello:
- 45502-84
- 45502-28
- 45502-85
- 45502-22
- 45502-80
- 45505-56
- 45502-56
- 45505-72
- Connection type (port 1):
- Female luer
- Female luer
- Female luer
- Female luer
- Female luer
- Male luer lock
- Female luer
- Male luer slip
- Connection type (port 2):
- Male luer slip
- Cap
- Female luer
- Female luer
- Male luer lock
- Plug
- Female luer
- Male luer slip
- Connection type (port 3):
- Male luer slip
- Female luer
- No. of ports:
- 2
- 1
- 2
- 2
- 2
- 1
- 3
- 2
- Garanzia:
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
Specifiche
Informazioni correlate
Frequently Bought Together