Hubtron™ Black Pascal Fluoroelastomer Tubing
Supplier: Avantor Fluid Handling
Certificates
About this item
Ideal for corrosive chemical transfer.
- Global market leader in range of formulations for most extensive chemical compatibility
Applications: Extremely demanding corrosive chemical transfer, electronics applications, ink and printing, and general manufacturing applications requiring UV protection.
Characteristics: Opaque black with UV block, extremely low gas permeability, extremely strong chemical resistance. Superior pressure ratings. Excellent chemical resistance to solvents, acids, bases, alcohols, and esters.
Certification: None.
Temperature: 14 to 302 °F (–5 to 150 °C)
Sterilization: Not recommended.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX95257-02
- MFLX95257-06
- MFLX95257-04
- Trasparenza:
- Opaque
- Opaque
- Opaque
- Length per pack:
- 7,6 m (25 ft.)
- 7,6 m (25 ft.)
- 7,6 m (25 ft.)
- Temperatura massima:
- 120 °C (248 °F)
- 120 °C (248 °F)
- 120 °C (248 °F)
- Temperatura minima:
- –10 °C (14 °F)
- –10 °C (14 °F)
- –10 °C (14 °F)
- Sterilisation method autoclave:
- No
- No
- No
- Sterilisation method EtO:
- No
- No
- No
- Sterilisation method gamma:
- No
- No
- No
- Garanzia:
- 1 Month
- 1 Month
- 1 Month