Masterflex® Transfer Tubing, Versilon™ 2001, Avantor®
Nome del fornitore: Avantor Fluid Handling
Certificati
Informazioni su questo articolo
Ultra-smooth inert inner bore provides excellent chemical resistance.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory approvals to match your unique application
This tubing is ideal for general laboratory use. They offer long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, are nontoxic. FDA compliant and made according to GMP. Temperature range is -51 to 165 °F (-46 to 73 °C). Can be sterilized with ethylene oxide or autoclave.
- Environmentally Preferable:
Specifiche
- Modello:
- 95716-06
- 95716-11
- 95716-14
- 95716-21
- 95716-29
- Trasparenza:
- Opaque
- Opaque
- Opaque
- Opaque
- Opaque
- Length per pack:
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- Pressione max.:
- 1,3 bar (19 psi)
- 0,6 bar (9 psi)
- 1,3 bar (19 psi)
- 0,6 bar (9 psi)
- 0,8 bar (12 psi)
- Temperatura minima:
- –57 °C (–71 °F)
- –57 °C (–71 °F)
- –57 °C (–71 °F)
- –57 °C (–71 °F)
- –57 °C (–71 °F)
- Temperatura massima:
- 73 °C (165 °F)
- 73 °C (165 °F)
- 73 °C (165 °F)
- 73 °C (165 °F)
- 73 °C (165 °F)
- Regulatory approvals:
- FDA
- FDA
- FDA
- FDA
- FDA
- Sterilisation method autoclave:
- Yes
- Yes
- Yes
- Yes
- Yes
- Sterilisation method EtO:
- Yes
- Yes
- Yes
- Yes
- Yes
- Sterilisation method gamma:
- Yes
- Yes
- Yes
- Yes
- Yes
- Pressione di esercizio:
- 1,3 bar (19 psi)
- 0,6 bar (9 psi)
- 1,3 bar (19 psi)
- 0,6 bar (9 psi)
- 0,8 bar (12 psi)