Masterflex® Ismatec® Minicartridge Pump Heads for Masterflex® L/S® Drives, Avantor®
Combine the accuracy of Ismatec® with the versatility of Masterflex® tubing pump systems.
- Order optional minicartridges with adjustable occlusion to vary pressure each channel
- Easily remove or insert minicartridges without disturbing other channels
- Max pressure of 1.5 bar is achieved with pressure-lever cassettes only
- Click-n-go cassettes offer a max pressure of 1.0 bar.
Choose from six, eight, or twelve roller heads. Less rollers offer greater flow rates, while more rollers offer a virtually pulseless flow. Heads accept up to four or eight Click’n’go™ minicartridges (included). Click’n’go minicartridges feature spring occlusion for optimum tension at all times with no adjustments.
Call our Application Specialists to determine the maximum recommended number of cartridges to use with your drive.
- Environmentally Preferable:
- Catalog No:
- MFLX07623-30
- MFLX07623-10
- MFLX07623-00
- Modello:
- 07623-30
- 07623-10
- 07623-00
- Max. flow rate per channel:
- 52 ml/min
- 156 ml/min
- 195 ml/min
- Min. flow rate per channel:
- 0,003 ml/min
- 0,002 ml/min
- 0,003 ml/min
- N. di canali:
- 4
- 8
- 8
- Occlusion:
- Adjustable
- Adjustable
- Adjustable
- Open-head sensor:
- No
- No
- No
- Optimum RPM range:
- 1 - 600 rpm
- 1 - 600 rpm
- 1 - 600 rpm
- Pump series:
- L/S
- L/S
- L/S
- Tubing retention:
- Stopped Tubing or Manual
- Stopped Tubing or Manual
- Stopped Tubing or Manual
- Tubing sizes accepted:
- Three-stop microbore
- Three-stop microbore
- Three-stop microbore
- Garanzia:
- 2 years
- 1 year
- 1 year
Frequently Bought Together