Orders LinkContactUsLinkComponent
Masterflex® Variable-Area Flowmeter Acrylic Tripod Bases, Avantor®
Masterflex® Variable-Area Flowmeter Acrylic Tripod Bases, Avantor®
Fournisseur:   Avantor Fluid Handling
Masterflex® Variable-Area Flowmeter Acrylic Tripod Bases, Avantor®
Fournisseur:  Avantor Fluid Handling
Certificats
Masterflex® Variable-Area Flowmeter Acrylic Tripod Bases, Avantor®
Masterflex® Variable-Area Flowmeter Acrylic Tripod Bases, Avantor®
3 variantes
Product Details & Documents
Easily mount flowmeters on tripod for easy viewing.
  • Constructed of durable acrylic
These tripod bases are ideal for benchtop applications. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting. Tripod bases are made of acrylic and include a spirit leveler.
Spécifications de la famille de produits
Spécifications
  • Modello:
  • 03218-58
  • 03226-40
  • 03226-50
  • Larghezza:
  • 1755"
  • 1755"
  • 1755"
  • Garanzia:
  • 1 year
  • 1 year
  • 1 year
Spécifications
Product Family Options
Informations sur le produit
  • Descrizione
    Housing material
    Disponibilité
    Prix
  • MFLX03218-58
    Tripod Base for 03218-50 and 03218-52
    Acrylic
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - SFr. 66.80
    SFr. 66.80
    1 * 1 ST
       SFr. 66.80
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MFLX03218-58
    Modello
    03218-58
    Larghezza
    1755"
    Garanzia
    1 year
  • MFLX03226-40
    Tripod Base for Three to Five Tube Systems
    Acrylic
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - SFr. 66.80
    SFr. 66.80
    1 * 1 ST
       SFr. 66.80
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MFLX03226-40
    Modello
    03226-40
    Larghezza
    1755"
    Garanzia
    1 year
  • MFLX03226-50
    Tripod Base for Multi-Tube Frames
    Acrylic
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - SFr. 66.80
    SFr. 66.80
    1 * 1 ST
       SFr. 66.80
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MFLX03226-50
    Modello
    03226-50
    Larghezza
    1755"
    Garanzia
    1 year