1536-well spheroid microplates, Corning®
Codice catalogo CORN4637
Nome del fornitore: Corning
Specifiche
- N. di pozzetti:1536
- Confezione:1 / Pk, 5 / Cs
- Volume di lavoro raccomandato:5 - 10 µl
- Sterili:+
- Colore:nero
- Descrizione:1536-well black/clear round bottom ultra-low attachment spheroid microplate, with lid, sterile
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:RECs, ISO 14001 - Kennebunk
- Conf:5
- Volume pozzetti:14 ml
Specifiche
Informazioni su questo articolo
With their proprietary design, these microplates are ideal for generating and analyzing 3D multicellular spheroids in the same microplate. The Ultra-Low Attachment (ULA) surface enables uniform and reproducible 3D multicellular spheroid formation. The black opaque microplate body shields each optically clear, round bottom well from well-to-well cross-talk.
- 536-well format allows for high throughput 3D cell culture
- Optically clear round bottom with black opaque microplate body
- Covalent attachment of Ultra-Low Attachment surface to reduce cellular adhesion to well surface
- Novel well geometry aids in the generation of uniform, single spheroids across all wells, which enables automated visualization
- Unique design shields each well to minimize well-to-well cross-talk
You can culture and assay spheroids in the same microplate, without the need for transfer to a new microplate. Compatible with the GNF ultra high throughput system.