pH/ion meter, SevenDirect SD50
Nome del fornitore: Mettler - Toledo
Informazioni su questo articolo
Robust bench top meter for accurate pH and ion determinations in the lab and near production.
- Large colour touch screen provides direct measurement and easy data entry
- Instructions guide users through the sensor calibration process
- Stores data, such as time stamp, sample and sensor ID with every measurement and prints or exports records automatically
- Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP 54 in-use)
The large colour touch screen offers direct menu access, fast data entry and guides users through routine tasks. The instruments supports GLP compliance by storing time stamp, sample, sensor and method IDs with every measurement
7 varianti
Product Details & Documents
Robust bench top meter for accurate pH and ion determinations in the lab and near production.
- Large colour touch screen provides direct measurement and easy data entry
- Instructions guide users through the sensor calibration process
- Stores data, such as time stamp, sample and sensor ID with every measurement and prints or exports records automatically
- Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP 54 in-use)
The large colour touch screen offers direct menu access, fast data entry and guides users through routine tasks. The instruments supports GLP compliance by storing time stamp, sample, sensor and method IDs with every measurement
Documents
Specifications for Product Family
Specifiche
- Catalog No:
- 662-2392
- 662-2384
- 662-2385
- 662-2386
- 662-2389
- 662-2387
- 662-2388
- pH - range:
- −2,000...+20,000
- −2,000...+20,000
- −2,000...+20,000
- −2,000...+20,000
- −2,000...+20,000
- −2,000...+20,000
- −2,000...+20,000
- pH - risoluzione:
- 0,1 / 0,01 / 0,001
- 0,1 / 0,01 / 0,001
- 0,1 / 0,01 / 0,001
- 0,1 / 0,01 / 0,001
- 0,1 / 0,01 / 0,001
- 0,1 / 0,01 / 0,001
- 0,1 / 0,01 / 0,001
- pH - accuratezza:
- ±0,002
- ±0,002
- ±0,002
- ±0,002
- ±0,002
- ±0,002
- ±0,002
- mV - range:
- −2000,0...+2000,0
- −2000,0...+2000,0
- −2000,0...+2000,0
- −2000,0...+2000,0
- −2000,0...+2000,0
- −2000,0...+2000,0
- −2000,0...+2000,0
- mV - risoluzione:
- 1 / 0,1
- 1 / 0,1
- 1 / 0,1
- 1 / 0,1
- 1 / 0,1
- 1 / 0,1
- 1 / 0,1
- mV - accuratezza:
- ±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
- ±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
- ±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
- ±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
- ±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
- ±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
- ±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
- ISE - gamma:
- mmol/L, mol/L, mg/L, ppm: 0,00000 - 999999 / %: 0,000 - 100,000 / pX: −2,000...20,000
- mmol/L, mol/L, mg/L, ppm: 0,00000 - 999999 / %: 0,000 - 100,000 / pX: −2,000...20,000
- mmol/L, mol/L, mg/L, ppm: 0,00000 - 999999 / %: 0,000 - 100,000 / pX: −2,000...20,000
- mmol/L, mol/L, mg/L, ppm: 0,00000 - 999999 / %: 0,000 - 100,000 / pX: −2,000...20,000
- mmol/L, mol/L, mg/L, ppm: 0,00000 - 999999 / %: 0,000 - 100,000 / pX: −2,000...20,000
- mmol/L, mol/L, mg/L, ppm: 0,00000 - 999999 / %: 0,000 - 100,000 / pX: −2,000...20,000
- mmol/L, mol/L, mg/L, ppm: 0,00000 - 999999 / %: 0,000 - 100,000 / pX: −2,000...20,000
- ISE risoluzione:
- mmol/L, mol/L, mg/L, ppm: 0,00001 / %: 0,001 / pX: 0,1 / 0,01
- mmol/L, mol/L, mg/L, ppm: 0,00001 / %: 0,001 / pX: 0,1 / 0,01
- mmol/L, mol/L, mg/L, ppm: 0,00001 / %: 0,001 / pX: 0,1 / 0,01
- mmol/L, mol/L, mg/L, ppm: 0,00001 / %: 0,001 / pX: 0,1 / 0,01
- mmol/L, mol/L, mg/L, ppm: 0,00001 / %: 0,001 / pX: 0,1 / 0,01
- mmol/L, mol/L, mg/L, ppm: 0,00001 / %: 0,001 / pX: 0,1 / 0,01
- mmol/L, mol/L, mg/L, ppm: 0,00001 / %: 0,001 / pX: 0,1 / 0,01
- ISE accuratezza:
- mmol/L, mol/L, mg/L, ppm, pX: ±0.002
- mmol/L, mol/L, mg/L, ppm, pX: ±0.002
- mmol/L, mol/L, mg/L, ppm, pX: ±0.002
- mmol/L, mol/L, mg/L, ppm, pX: ±0.002
- mmol/L, mol/L, mg/L, ppm, pX: ±0.002
- mmol/L, mol/L, mg/L, ppm, pX: ±0.002
- mmol/L, mol/L, mg/L, ppm, pX: ±0.002
- Range di temperatura:
- −30,0…+130,0 °C
- −30,0…+130,0 °C
- −30,0…+130,0 °C
- −30,0…+130,0 °C
- −30,0…+130,0 °C
- −30,0…+130,0 °C
- −30,0…+130,0 °C
- Risoluzione della temperatura:
- 0,1 °C
- 0,1 °C
- 0,1 °C
- 0,1 °C
- 0,1 °C
- 0,1 °C
- 0,1 °C
- Accuratezza della temperatura:
- ±0,1 (0...100 °C)
- ±0,1 (0...100 °C)
- ±0,1 (0...100 °C)
- ±0,1 (0...100 °C)
- ±0,1 (0...100 °C)
- ±0,1 (0...100 °C)
- ±0,1 (0...100 °C)
- Calibrazione:
- Linear, segmented
- Linear, segmented
- Linear, segmented
- Linear, segmented
- Linear, segmented
- Linear, segmented
- Linear, segmented
- Punti di calibrazione:
- 1 - 5
- 1 - 5
- 1 - 5
- 1 - 5
- 1 - 5
- 1 - 5
- 1 - 5
Specifiche
Product Family Options
Informazioni sul prodotto
- DisponibilitàPrice
- 662-2392pH/Ion meter, SevenDirect SD50Specifiche:
More
Specifications
Cat. No.662-2392pH - range−2,000...+20,000pH - risoluzione0,1 / 0,01 / 0,001pH - accuratezza±0,002mV - range−2000,0...+2000,0mV - risoluzione1 / 0,1mV - accuratezza±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)ISE - gammammol/L, mol/L, mg/L, ppm: 0,00000 - 999999 / %: 0,000 - 100,000 / pX: −2,000...20,000ISE risoluzionemmol/L, mol/L, mg/L, ppm: 0,00001 / %: 0,001 / pX: 0,1 / 0,01ISE accuratezzammol/L, mol/L, mg/L, ppm, pX: ±0.002Range di temperatura−30,0…+130,0 °CRisoluzione della temperatura0,1 °CAccuratezza della temperatura±0,1 (0...100 °C)CalibrazioneLinear, segmentedPunti di calibrazione1 - 5 - 662-2384pH/Ion meter, SevenDirect SD50 kitSpecifiche:
More
Specifications
Cat. No.662-2384pH - range−2,000...+20,000pH - risoluzione0,1 / 0,01 / 0,001pH - accuratezza±0,002mV - range−2000,0...+2000,0mV - risoluzione1 / 0,1mV - accuratezza±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)ISE - gammammol/L, mol/L, mg/L, ppm: 0,00000 - 999999 / %: 0,000 - 100,000 / pX: −2,000...20,000ISE risoluzionemmol/L, mol/L, mg/L, ppm: 0,00001 / %: 0,001 / pX: 0,1 / 0,01ISE accuratezzammol/L, mol/L, mg/L, ppm, pX: ±0.002Range di temperatura−30,0…+130,0 °CRisoluzione della temperatura0,1 °CAccuratezza della temperatura±0,1 (0...100 °C)CalibrazioneLinear, segmentedPunti di calibrazione1 - 5 - 662-2385pH/Ion meter, SevenDirect SD50 F-Ion kitSpecifiche:
More
Specifications
Cat. No.662-2385pH - range−2,000...+20,000pH - risoluzione0,1 / 0,01 / 0,001pH - accuratezza±0,002mV - range−2000,0...+2000,0mV - risoluzione1 / 0,1mV - accuratezza±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)ISE - gammammol/L, mol/L, mg/L, ppm: 0,00000 - 999999 / %: 0,000 - 100,000 / pX: −2,000...20,000ISE risoluzionemmol/L, mol/L, mg/L, ppm: 0,00001 / %: 0,001 / pX: 0,1 / 0,01ISE accuratezzammol/L, mol/L, mg/L, ppm, pX: ±0.002Range di temperatura−30,0…+130,0 °CRisoluzione della temperatura0,1 °CAccuratezza della temperatura±0,1 (0...100 °C)CalibrazioneLinear, segmentedPunti di calibrazione1 - 5 - 662-2386pH/Ion meter, SevenDirect SD50 HA kitSpecifiche:
More
Specifications
Cat. No.662-2386pH - range−2,000...+20,000pH - risoluzione0,1 / 0,01 / 0,001pH - accuratezza±0,002mV - range−2000,0...+2000,0mV - risoluzione1 / 0,1mV - accuratezza±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)ISE - gammammol/L, mol/L, mg/L, ppm: 0,00000 - 999999 / %: 0,000 - 100,000 / pX: −2,000...20,000ISE risoluzionemmol/L, mol/L, mg/L, ppm: 0,00001 / %: 0,001 / pX: 0,1 / 0,01ISE accuratezzammol/L, mol/L, mg/L, ppm, pX: ±0.002Range di temperatura−30,0…+130,0 °CRisoluzione della temperatura0,1 °CAccuratezza della temperatura±0,1 (0...100 °C)CalibrazioneLinear, segmentedPunti di calibrazione1 - 5 - 662-2389pH/Ion meter, SevenDirect SD50 LowVol kitSpecifiche:
More
Specifications
Cat. No.662-2389pH - range−2,000...+20,000pH - risoluzione0,1 / 0,01 / 0,001pH - accuratezza±0,002mV - range−2000,0...+2000,0mV - risoluzione1 / 0,1mV - accuratezza±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)ISE - gammammol/L, mol/L, mg/L, ppm: 0,00000 - 999999 / %: 0,000 - 100,000 / pX: −2,000...20,000ISE risoluzionemmol/L, mol/L, mg/L, ppm: 0,00001 / %: 0,001 / pX: 0,1 / 0,01ISE accuratezzammol/L, mol/L, mg/L, ppm, pX: ±0.002Range di temperatura−30,0…+130,0 °CRisoluzione della temperatura0,1 °CAccuratezza della temperatura±0,1 (0...100 °C)CalibrazioneLinear, segmentedPunti di calibrazione1 - 5 - 662-2387pH/Ion meter, SevenDirect SD50 OrganicSpecifiche:
More
Specifications
Cat. No.662-2387pH - range−2,000...+20,000pH - risoluzione0,1 / 0,01 / 0,001pH - accuratezza±0,002mV - range−2000,0...+2000,0mV - risoluzione1 / 0,1mV - accuratezza±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)ISE - gammammol/L, mol/L, mg/L, ppm: 0,00000 - 999999 / %: 0,000 - 100,000 / pX: −2,000...20,000ISE risoluzionemmol/L, mol/L, mg/L, ppm: 0,00001 / %: 0,001 / pX: 0,1 / 0,01ISE accuratezzammol/L, mol/L, mg/L, ppm, pX: ±0.002Range di temperatura−30,0…+130,0 °CRisoluzione della temperatura0,1 °CAccuratezza della temperatura±0,1 (0...100 °C)CalibrazioneLinear, segmentedPunti di calibrazione1 - 5 - 662-2388pH/Ion meter, SevenDirect SD50 Pure H₂OSpecifiche:
More
Specifications
Cat. No.662-2388pH - range−2,000...+20,000pH - risoluzione0,1 / 0,01 / 0,001pH - accuratezza±0,002mV - range−2000,0...+2000,0mV - risoluzione1 / 0,1mV - accuratezza±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)ISE - gammammol/L, mol/L, mg/L, ppm: 0,00000 - 999999 / %: 0,000 - 100,000 / pX: −2,000...20,000ISE risoluzionemmol/L, mol/L, mg/L, ppm: 0,00001 / %: 0,001 / pX: 0,1 / 0,01ISE accuratezzammol/L, mol/L, mg/L, ppm, pX: ±0.002Range di temperatura−30,0…+130,0 °CRisoluzione della temperatura0,1 °CAccuratezza della temperatura±0,1 (0...100 °C)CalibrazioneLinear, segmentedPunti di calibrazione1 - 5
Accessories
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Parking beaker insert, EasyPlace; should be used while using the parking beaker with storage solutionParking beaker insert, EasyPlace; should be used while using the parking beaker with storage solution662-2416Log in per disponibilità1 * 1 ST - SFr. 9.80
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Parking beaker, EasyPlace electrode arm; Fill it with storage solution and keep your electrode perfectly conditioned also in short breaks between measurements; or use it to store the wetting cap while measuringParking beaker, EasyPlace electrode arm; Fill it with storage solution and keep your electrode perfectly conditioned also in short breaks between measurements; or use it to store the wetting cap while measuring662-2415Log in per disponibilità1 * 1 ST - SFr. 26.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Sensor holder, EasyPlace Single for the EasyPlace electrode arm holds one sensorSensor holder, EasyPlace Single for the EasyPlace electrode arm holds one sensor662-2418Log in per disponibilità1 * 1 ST - SFr. 26.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Sachet holder, EasyPlace rotating; hold up to three buffer sachets and prevent spillage; It can be rotated to place one buffer after the other in the calibration positionSachet holder, EasyPlace rotating; hold up to three buffer sachets and prevent spillage; It can be rotated to place one buffer after the other in the calibration position662-2417Log in per disponibilità1 * 1 ST - SFr. 50.30
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Sensor Holder, EasyPlace Triple for the EasyPlace electrode arm holds up to three sensorsSensor Holder, EasyPlace Triple for the EasyPlace electrode arm holds up to three sensors662-2419Log in per disponibilità1 * 1 ST - SFr. 31.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Pole EasyPlace, 30 cm, standard pole for the EasyPlace electrode armPole EasyPlace, 30 cm, standard pole for the EasyPlace electrode arm662-2421Log in per disponibilità1 * 1 ST - SFr. 251.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Sensor Holder, EasyPlace Single Extended for the EasyPlace electrode arm with extended length for use with large sample vessels. Holds one sensorSensor Holder, EasyPlace Single Extended for the EasyPlace electrode arm with extended length for use with large sample vessels. Holds one sensor662-2420Log in per disponibilità1 * 1 ST - SFr. 31.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Pole, EasyPlace 50 cm, extended pole for the EasyPlace electrode arm for use with large sample vesselsPole, EasyPlace 50 cm, extended pole for the EasyPlace electrode arm for use with large sample vessels662-2422Log in per disponibilità1 * 1 ST - SFr. 302.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Base plate for the EasyPlace electrode arm
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Power supply, 18 W, does not include country-specific plug (order plugs separately with model (662-2426)Power supply, 18 W, does not include country-specific plug (order plugs separately with model (662-2426)662-2424Log in per disponibilità1 * 1 ST - SFr. 100.50
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Top cover pole, EasyPlace or the top of the EasyPlace pole, has two positions for long-term sensor storageTop cover pole, EasyPlace or the top of the EasyPlace pole, has two positions for long-term sensor storage662-2425Log in per disponibilità1 * 1 ST - SFr. 26.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Plug set, set of country-specific plugs for Power supply 18 W (662-2424)Plug set, set of country-specific plugs for Power supply 18 W (662-2424)662-2426Log in per disponibilità1 * 1 ST - SFr. 34.10
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Replaceable protective cover for SevenDirect metersReplaceable protective cover for SevenDirect meters662-2382Log in per disponibilità1 * 1 ST - SFr. 58.30
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Screws and feet set EasyPlace
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Electrode arm, EasyPlace Complete supports efficient measurement and calibration; With perfect vertical movement, the risk of any spillage is minimised; Includes parking beaker, parking beaker insert and sachet holderElectrode arm, EasyPlace Complete supports efficient measurement and calibration; With perfect vertical movement, the risk of any spillage is minimised; Includes parking beaker, parking beaker insert and sachet holder662-2413Log in per disponibilità1 * 1 ST - SFr. 352.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Magnetic stirrer, EasyMix for use with SevenDirect meters; The EasyMix can be controlled manually or automatically by the SevenDirect meterMagnetic stirrer, EasyMix for use with SevenDirect meters; The EasyMix can be controlled manually or automatically by the SevenDirect meter662-2414Log in per disponibilità1 * 1 ST - SFr. 417.00