Orders LinkContactUsLinkComponent
Bonn micro forceps
Bonn micro forceps
Marketsource
Supplier:   FINE SCIENCE TOOLS
Bonn micro forceps
Supplier:  FINE SCIENCE TOOLS
Marketsource
Certificates
Bonn micro forceps
Bonn micro forceps
2 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Materiale
    Lunghezza
    Stile punta
    Tip shape
    Tip size
    Tip width
    Availability
    Price
  • Acciaio inossidabile
    70 mm
    1×2 teeth
    Straight
    0,3×0,25 mm
    0,45 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 229.94
    SFr. 229.94
    1 * 1 ST
       SFr. 229.94
    Specifications:
    Marketsource
    Marketsource
  • Acciaio inossidabile
    70 mm
    Smooth
    Straight
    0,3×0,25 mm
    0,3 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 180.47
    SFr. 180.47
    1 * 1 ST
       SFr. 180.47
    Specifications:
    Marketsource
    Marketsource