Dumont #2, laminectomy forceps
#2, Inox, 120 mm, Standard, Straight, 0,4×0,02 mm
FSCI11223-20
Marketsource
- Tipo:#2
- Lunghezza:120 mm
- Materiale:Inox
- Stile punta:Standard
- Tip size:0,4×0,02 mm
- Tip shape:Straight