Dumont #L5, sliding lock forceps
Catalog # FSCI11253-27
Supplier: FINE SCIENCE TOOLS
Specifications
- Tipo:#L5
- Lunghezza:110 mm
- Materiale:Inox
- Stile punta:Standard
- Conf:1
- Tip size:0,1×0,06 mm
- Tip shape:Straight
Specifications
About this item
Plastic clamping ring holds tips together and can be locked in place. Fully autoclavable.