Orders LinkContactUsLinkComponent
Moria Iris forceps
Marketsource
Moria Iris forceps
Marketsource
Moria Iris forceps
Moria Iris forceps
Product Family Options
  • Tipo
    Materiale
    Lunghezza
    Stile punta
    Tip shape
    Tip size
    Tip width
  • MC31
    Inox
    100 mm
    Serrated
    Curved
    0,5×0,4 mm
    0,5 mm
    1 * 1 ST
    1 * 1 ST - SFr. 139.71
    SFr. 139.71
    1 * 1 ST
       SFr. 139.71
    Marketsource
    Marketsource
  • MC32
    Inox
    100 mm
    Serrated
    Straight
    0,5×0,3 mm
    0,5 mm
    1 * 1 ST
    1 * 1 ST - SFr. 134.67
    SFr. 134.67
    1 * 1 ST
       SFr. 134.67
    Marketsource
    Marketsource
  • MC31B
    Inox
    100 mm
    Smooth
    Curved
    0,5×,04 mm
    0,5 mm
    1 * 1 ST
    1 * 1 ST - SFr. 132.83
    SFr. 132.83
    1 * 1 ST
       SFr. 132.83
    Marketsource
    Marketsource
  • MC32B
    Inox
    100 mm
    Smooth
    Straight
    0,5×0,3 mm
    0,5 mm
    1 * 1 ST
    1 * 1 ST - SFr. 128.71
    SFr. 128.71
    1 * 1 ST
       SFr. 128.71