Orders LinkContactUsLinkComponent
Halsey needle holders
Marketsource
Halsey needle holders
Marketsource
Halsey needle holders
Halsey needle holders
Product Family Options
  • Descrizione
    Tipo
    Materiale
    Lunghezza
    Stile punta
    Tip shape
    Tip width
  • Clamping length of 16 mm
    Lockable
    Acciaio inossidabile
    130 mm
    Straight
    1,9 mm
    1 * 1 ST
    1 * 1 ST - 31.61 SFr.
    31.61 SFr.
    1 * 1 ST
       31.61 SFr.
    Marketsource
    Marketsource
  • Clamping length of 15 mm
    Lockable
    Acciaio inossidabile
    130 mm
    Serrated, tungsten carbide
    Straight
    2 mm
    1 * 1 ST
    1 * 1 ST - 100.31 SFr.
    100.31 SFr.
    1 * 1 ST
       100.31 SFr.