Silver neuro clips
Silver Neuro Clips, 5 mm
Catalog # FSCI18040-00
Marketsource
Supplier: FINE SCIENCE TOOLS
Jump to:
Product Details & Documents
These single-use silver neuro clips are meant for permanent vessel occlusion.
- Clamp Press: Variable
- Alloy / Material: Silver
- Length: 5 mm
- Not serrated
Caution: All surgical instruments and devices from the Fine Science Tools product range are intended exclusively for use in experimental research laboratories and facilities or in veterinary medicine.
Specifications
- Descrizione:Silver Neuro Clips
- Taglia:5 mm