Beyer rongeur
Beyer rongeurs, 2,5 mm cup diameter, Curved, 180 mm
Codice catalogo FSCI16002-18
Marketsource
Nome del fornitore: FINE SCIENCE TOOLS
Vai a
Product Details & Documents
These rongeurs are made of stainless steel.
- Double joint action
Specifiche
- Tipo:Curved
- Descrizione:Beyer rongeurs, 2,5 mm cup diameter
- Lunghezza:180 mm