Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Forbici
Lister scissors
Lister scissors
FSCI14131-14
:
FINE SCIENCE TOOLS
Lister scissors
FSCI14131-14
:
FINE SCIENCE TOOLS
Tipo:
Blunt/blunt, angled to side tips
Lunghezza:
140 mm
Conf:
1
Serrated:
+
Lunghezza lama:
33 mm
These scissors are made of stainless steel.