ContactUsLinkComponent
Dissector scissors, slim blades
Catalog # FSCI14081-09
Anbieter:  FINE SCIENCE TOOLS
undefined
Dissector scissors, slim blades
Catalog # FSCI14081-09
Anbieter:  FINE SCIENCE TOOLS
Technische Daten
  • Tipo:
    Sharp/sharp, angled to side tips
  • Lunghezza:
    100 mm
  • Conf:
    1
  • Lunghezza lama:
    8 mm
Technische Daten
Über diesen Artikel
These scissors are made of stainless steel.