Polycarbonate retractable eyeshield, EVOSpec®
EVOSpec® retractable eyeshield for assembly to helmets, clear PC
Codice catalogo JSPAANT010-200-000
Nome del fornitore: JSP
Polycarbonate retractable eyeshield, EVOSpec®
Codice catalogo JSPAANT010-200-000
Nome del fornitore: JSP
Vai a
Product Details & Documents
Easy assembly kit for adding a clear, polycarbonate, retractable eyeshield to existing Evolution® helmets.
- Polycarbonate retractable eye shield
- 6 Base curve lens offers optimum coverage for all users
- Low profile bi-polymer nose clip for improved comfort and security
- Factory fit and/or user replaceable
- Polypropylene arms
- Anti-fog and anti-scratch treatment
Stylish design, retracts neatly and securely into helmet when not in use. Compatible with Evolution® safety helmets (EVOLite® / EVO3®/EVO5®).
Certifications: Conforms to EN 166.1.F.
Documents
TDS Spectacles Evospec Clear
Scheda dati
Specifiche
- Descrizione:EVOSpec® retractable eyeshield for assembly to helmets, clear PC