Quick release, 4-point Linesman chinstrap harness
Catalog # JSPAAHV200-000-000
Supplier: JSP
Specifications
- Descrizione:Quick Release, 4-point linesman chinstrap harness
- Conf:1
Specifications
About this item
The JSP 4-point Linesman chinstrap provides increased security for a firmer fit, preventing helmets being accidentally dropped during extreme movement.
- 4-point chinstrap for Evolution® safety helmets
- Tubular nylon webbing
- LDPE clip
- Nylon buckle
- Increased security for a firmer fit
The JSP 4-point Linesman chinstrap provides increased security for a firmer fit, preventing helmets from being accidentally dropped during extreme movement. Simple, secure closure from the nylon buckle allows quick donning and doffing of the helmet.
Certifications: EN 397 EN 14052. Meets the requirements of EN 397 and EN 14052 when worn with the appropriate JSP EVO® helmet.