Junior IV goggle
Junior IV goggle, Polycarbonate
Katalognr. JSPAAGT020-141-300
Anbieter: JSP
Direkt zu
Product Details & Documents
Junior goggle with reduced size housing, fully conforming to EN 166 34B.
- Junior goggle
- Reduced size housing
- One-piece polycarbonate lens
- Elastic headband
The lens and body offer protection against medium energy, medium-speed particles at 120 m/s, large dust particles, and liquid droplets. The optical qualities of the lens comply with the requirements of optical class 1 (highest).
Zertifizierung: EN 166, EN 170.
Dokumente
TDS Goggles Junior Iv
Technisches Datenblatt
Technische Daten
- Descrizione:Junior IV goggle
- Materiale lente:Polycarbonate