Orders LinkContactUsLinkComponent
Isolated amplifier, b.safe
Isolated amplifier, ABS, 160×60×160 mm
  BOHLR660-01
 :   Bohlender
Isolated amplifier, b.safe
  BOHLR660-01
 :  Bohlender
Isolated amplifier, b.safe
Isolated amplifier, b.safe
Isolated amplifier, ABS, 160×60×160 mm
SFr. 805.00
1 * 1 ST
Enclosed housing for installation at the rear side of a safety cabinet. Unit provides safe signal exchange between explosion-hazardous and explosion-proof areas by galvanic isolation of the sensor side and the output side.
  • For galvanic isolation of the sensor side and the output side
  • Ignitable sparks are prevented in the explosive atmosphere on the sensor side
Ready for installation plastic housing made of ABS (IP 66 rated) that prevents accidental contact of live parts. With clearly labelled housing screw connections to connect up to two b.safe Filling Level Sensors and to operate them safely in explosion-proof areas. Includes power supply (90 to 264 V / 47 to 63 Hz) with plug adapter for EU, Australia, USA, Japan, and UK. The installation of b.safe Isolated Amplifier and the connection of the signal cable must be carried out by qualified personnel.
By using the b.safe Isolated Amplifier in an explosion-proof area, the electronic alerting system prevents ignitable sparks from arising in the explosive atmosphere around the sensors. Combined with the b.safe Display, the b.safe Isolated Amplifier enables an automatic recognition of damaged property, disconnected sensors, or line breaks. If the b.safe Isolated Amplifier can be used with filling level monitoring devices of other brands, it must be checked individually.
  • Descrizione:
    Isolated amplifier, ABS
  • L×P×A:
    160×60×160 mm