Orders LinkContactUsLinkComponent
Writable Cryo Colour Rectangles and Dots
Nome del fornitore:   GA
Writable Cryo Colour Rectangles and Dots
Nome del fornitore:  GA
Certificates
Writable Cryo Colour Rectangles and Dots
Writable Cryo Colour Rectangles and Dots
61 varianti
Product Details & Documents
Writable cryogenic colour dots and rectangles provided in dispenser box rolls for identifying the sides and tops of cryo vials and microtubes stored in lab freezers and liquid nitrogen dewars. Easy to write on with permanent cryo markers, JTRA-class labels are available in a variety of colours.
  • Temperature range: −196 to +110 °C (−320.8 to +230 °F)
  • Designed for use in liquid nitrogen (−196 °C), freezers (−80 °C), and dry ice
  • Ideal for eppendorf microtubes and cryo vials
  • Strong adhesive ensures labels won't wrinkle or peel during long-term storage
  • Available in a wide range of colours, for color-coding and easy identification
These writable cryogenic rectangle and dot labels for cryo vials and conical bottom microtubes are made with a strong permanent adhesive ideal for liquid nitrogen, lab freezers, and dry ice storage and transportation. Easily writable with permanent cryogenic markers, these ultra-low temperature labels are supplied in roll format and are available in 15 individual colours as well as packs of 15 assorted colours for colour coding and rapid identification.
Product Family Options
Circle permanent cryogenic writable labels
Informazioni sul prodotto
  • Colore
    Dimensioni
    Confezione
    Disponibilità
    Price
  • GAINJTRA-9BE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    beige
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-9BL
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    blu
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-9BR
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    marrone
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-9GA
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    verde
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-9GS
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    verde chiaro
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-9LA
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    porpora chiaro
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-9LE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    giallo chiaro
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-9MT
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Verde menta
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-9OR
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    arancione
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-9PI
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    rosa
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-9RE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    rosso
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-9SI
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    argento
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-9VI
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    viola
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-9WH
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    bianco
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-9YE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    giallo
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-11BE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    beige
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 420 ROU
    1 * 420 ROU - SFr. 38.20
    SFr. 38.20
    1 * 420 ROU
       SFr. 38.20
  • GAINJTRA-11BL
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    blu
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-11BR
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    marrone
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-11GA
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    verde
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-11GS
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    verde chiaro
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-11LA
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    porpora chiaro
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-11LE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    giallo chiaro
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-11MT
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Verde menta
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-11OR
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    arancione
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-11PI
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    rosa
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-11RE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    rosso
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-11SI
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    argento
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-11VI
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    viola
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-11WH
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    bianco
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-11YE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    giallo
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-13BE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    beige
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-13BL
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    blu
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-13BR
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    marrone
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-13GA
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    verde
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-13GS
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    verde chiaro
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-13LA
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    porpora chiaro
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-13LE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    giallo chiaro
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-13MT
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Verde menta
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-13OR
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    arancione
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-13PI
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    rosa
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-13RE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    rosso
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-13SI
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    argento
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-13VI
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    viola
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-13WH
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    bianco
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-13X6A
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Multicolore
    12,7 mm (0,5")
    5040 labels/roll (6 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 156.00
    SFr. 156.00
    1 * 1 ROU
       SFr. 156.00
  • GAINJTRA-13YE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    giallo
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
Rectangle permanent cryogenic writable labels
Informazioni sul prodotto
  • Colore
    Dimensioni
    Confezione
    Disponibilità
    Price
  • GAINJTRA-2610BE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    beige
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-2610BL
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    blu
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-2610BR
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    marrone
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-2610GA
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    verde
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-2610GS
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    verde chiaro
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-2610LA
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    porpora chiaro
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-2610LE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    giallo chiaro
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-2610MT
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Verde menta
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-2610OR
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    arancione
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-2610PI
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    rosa
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-2610RE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    rosso
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-2610SI
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    argento
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-2610VI
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    viola
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-2610WH
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    bianco
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20
  • GAINJTRA-2610YE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    giallo
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ROU
    1 * 1 ROU - SFr. 38.20
    SFr. 38.20
    1 * 1 ROU
       SFr. 38.20