Angled connector, b.safe
Angled connector, PE, 90°, 1× clamp connection Ø 32 mm and 1× GL25 male
Codice catalogo BOHLR1185-01
Nome del fornitore: Bohlender
Vai a
Product Details & Documents
Angular piece approx. 90° made of static dissipative PE-EX.
Connection 1: Lock nut and clamp ring for inserting and clamping b.safe Waste Pipes with Ext.Ø 32 mm.
Connection 2: Male thread GL 25 for transition to flexible b.safe Waste tubes.
Specifiche
- Descrizione:Angled connector, PE, 90°, 1× clamp connection Ø 32 mm and 1× GL25 male