ContactUsLinkComponent
Connector for waste pipes, b.safe
Connector for waste pipes, b.safe
Catalog # BOHLR1172-01
Supplier:  Bohlender
undefined
Connector for waste pipes, b.safe
Catalog # BOHLR1172-01
Supplier:  Bohlender
Specifications
  • Descrizione:
    Connector for waste pipes, PE, 2× clamp connection Ø 32 mm
  • Conf:
    1
Specifications
About this item
Straight connector made of static dissipative PE-EX.
With lock nuts and clamp rings on both sides for fixing and clamping b.safe waste pipes with Ext.Ø 32 mm.