DYMO-compatible cryogenic labels
DYMO compatible, direct thermal cryo labels. For ultra-low and cryogenic temperature applications as low as –196 °C.
- EG1F-081 series labels are resistant to gamma irradiation (tested to up to 50 kGy)
- Ideal for the identification of 0,5 to 5 ml vials and tubes
- Compatible with all DYMO label writers
- Temperature range: –196 to 70 °C
- Permanent adhesive
Ideal for identification and permanent storage of vials and containers in liquid nitrogen and laboratory freezers (–80, –50, –20 °C). Labels will also resist extreme cryogenic temperatures (–196 °C in a liquid phase and vapor phase liquid nitrogen).
DYMO compatible, direct thermal cryo labels. For ultra-low and cryogenic temperature applications as low as –196 °C.
- EG1F-081 series labels are resistant to gamma irradiation (tested to up to 50 kGy)
- Ideal for the identification of 0,5 to 5 ml vials and tubes
- Compatible with all DYMO label writers
- Temperature range: –196 to 70 °C
- Permanent adhesive
Ideal for identification and permanent storage of vials and containers in liquid nitrogen and laboratory freezers (–80, –50, –20 °C). Labels will also resist extreme cryogenic temperatures (–196 °C in a liquid phase and vapor phase liquid nitrogen).
Product Family Options
- DescrizioneColoreDimensioni
- Rectangle cryo labelsbeige12,7×26×9,5 mm1 * 1 ROUSFr. 108.00
- Rectangle cryo labelsblu12,7×26×9,5 mm1 * 1 ROUSFr. 108.00
- Rectangle cryo labelsarancione12,7×26×9,5 mm1 * 1 ROUSFr. 108.00
- Rectangle cryo labelsrosso12,7×26×9,5 mm1 * 1 ROUSFr. 108.00
- Rectangle cryo labelsviola12,7×26×9,5 mm1 * 1 ROUSFr. 108.00
- Rectangle cryo labelsgiallo12,7×26×9,5 mm1 * 1 ROUSFr. 108.00
- Round cryo labelsbeige26,4×12,7×9,5 mm circle1 * 1 ROUSFr. 108.00
- Round cryo labelsblu26,4×12,7×9,5 mm circle1 * 1 ROUSFr. 108.00
- Round cryo labelsmarrone26,4×12,7×9,5 mm circle1 * 1 ROUSFr. 108.00
- Rectangle labels plus round cryo tagsverde26,4×12,7×9,5 mm circle1 * 1 ROUSFr. 108.00
- Rectangle labels plus round cryo tagsverde26,4×12,7×9,5 mm circle1 * 1 ROUSFr. 108.00
- Rectangle labels plus round cryo tagsgiallo26,4×12,7×9,5 mm circle1 * 1 ROUSFr. 108.00
- Rectangle labels plus round cryo tagsarancione26,4×12,7×9,5 mm circle1 * 1 ROUSFr. 108.00
- Round cryo labelsrosso26,4×12,7×9,5 mm circle1 * 1 ROUSFr. 108.00
- Round cryo labelsviola26,4×12,7×9,5 mm circle1 * 1 ROUSFr. 108.00
- Round cryo labelsgiallo26,4×12,7×9,5 mm circle1 * 1 ROUSFr. 108.00
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..