Face shield visor, 701.02
Interchangeable visor, Anti-fog, Acetato
Codice catalogo UNVT1220010
Nome del fornitore: UNIVET
Vai a
Product Details & Documents
The 701.02 face shield meets rigorous EN 166 standards (droplets and splashes) by providing visor with extended coverage on the upper part of the forehead.
- Anti-fog acetate visor
- Adjustable elastic head band
- Visor inclines up to 90°
- Interchangeable visor
- Face shield suitable for use in combination with prescription glasses and protective masks
This anti-fog visor allows maximum vision due to anti-fog characteristics. A soft sponge in front with an easily adjustable band ensures comfort. This visor is suitable for use with corrective glasses and respiratory protective masks.
Certifications: Certified according to EN 166, marking 3 protection against droplets and liquid splash.
Specifiche
- Tipo:Anti-fog
- Descrizione:Interchangeable visor
- Materiale:Acetato