AssetTag
AssetTag, 40×90 mm (W×H)
Codice catalogo SENA0096647494982
Nome del fornitore: SenseAnywhere
Vai a
Product Details & Documents
An AssetTag is a card with barcode for the holder and QR code for on the outside of an asset representing the monitored asset in SAClient.
- Monitor temp/RH of the asset
- Data loggers can be switched at any time
- Complete temp/RH history of assets without any monitoring-gaps
- No reconfiguration needed (alarms, reports etc.)
- No paperwork or administration
- Not prone to error or hassle
- Complete audit trail
How it works: The back of the card has two-sided foam tape. The card can be snapped into two parts. Bottom part: attach to holder in the asset. Top part: on outside of the asset (fridge/freezer). Scan the QR code to see the state of the asset. See which device (serial number) is attached. Proceed to SAClient for coupling, decoupling or swapping.
Documents
TDS AssetTag 20-00-00
Scheda dati
Specifiche
- Descrizione:AssetTag, 40×90 mm (W×H)