Orders LinkContactUsLinkComponent
Student Vannas spring scissors
Student Vannas spring scissors
Marketsource
Supplier:   FINE SCIENCE TOOLS
Student Vannas spring scissors
Supplier:  FINE SCIENCE TOOLS
Marketsource
Certificates
Student Vannas spring scissors
Student Vannas spring scissors
2 Options Available Below
Product Details & Documents
These scissors are made of stainless steel.
Designed with both quality and economy in mind, the student quality instruments are German made, but they have less hand finishing and use less expensive alloys so they can be offered at a considerably lower price.
Product Family Options
Product Information
  • Tipo
    Lunghezza
    Lunghezza lama
    Tip size
    Availability
    Price
  • Student quality, sharp, curved tips
    90 mm
    5 mm
    0,35 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 63.67
    SFr. 63.67
    1 * 1 ST
       SFr. 63.67
    Specifications:
    Marketsource
    Marketsource
  • Student quality, sharp, straight tips
    90 mm
    5 mm
    0,35 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 61.38
    SFr. 61.38
    1 * 1 ST
       SFr. 61.38
    Specifications:
    Marketsource
    Marketsource