Anti-CD193 (CCR3) Rat Monoclonal Antibody [clone: J073E5] (APC (Allophycocyanin))
Nome del fornitore: Biolegend
Certificati
Informazioni su questo articolo
Specifiche
- Catalog No:
- BLEG144511
- BLEG144512
- Simbolo dell'antigene:
- CD193 (CCR3)
- CD193 (CCR3)
- Sinomini antigene:
- eotaxin receptor|MIP1-alpha receptor like-2|CC CKR3|MIP1-α receptor like-2
- eotaxin receptor|MIP1-alpha receptor like-2|CC CKR3|MIP1-α receptor like-2
- Clonalità:
- Monoclonale
- Monoclonale
- Coniugazione:
- APC (Allofilcocianina)
- APC (Allofilcocianina)
- Clone:
- J073E5
- J073E5
- Reattività:
- Mouse
- Mouse
- Ospite:
- Rat
- Rat
- Isotipo:
- IgG2a kappa
- IgG2a kappa
- Citometria a flusso:
- Yes
- Yes
- Immunogeno:
- Mouse CCR3-transfectants
- Mouse CCR3-transfectants
- Purificazione:
- The antibody was purified by affinity chromatography and conjugated with APC under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with APC under optimal conditions.
- Concentrazione:
- 0.2 mg/ml
- 0.2 mg/ml
- Tampone di conservazione:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Temperatura di conservazione:
- 2…8 °C
- 2…8 °C
- Applicazioni testate:
- FC
- FC
Specifiche
Frequently Bought Together