Anti-Asialo-GM1 Rabbit Polyclonal Antibody [clone: Poly21460] (Ultra-LEAF™ )
Catalog # BLEG146002
Supplier: Biolegend
Specifications
- Simbolo dell'antigene:Asialo-GM1
- Clonalità:Policlonale
- Clone:Poly21460
- Coniugazione:Ultra-LEAF™
- Citometria a flusso:Yes
- Ospite:Rabbit
- Isotipo:IgG
- Reattività:Rat,Mouse
- Sinomini antigene:GM1|Ganglioside|ganglio-N-tetraosylceramide
- Tampone di conservazione:Lyophilized powder. Endotoxin level is <0.01 EU/µg of the protein (<0.001 ng/µg of the protein) as determined by the LAL test.
- Temperatura di conservazione:2…8 °C
- Immunogeno:Asialo-GM1
- Applicazioni testate:FC, Depletion
- Purificazione:Reconstitute with 1 ml sterile deionized, distilled, endotoxin-free water. Gamma globulin fraction of serum was precipitated using 50% ammonium sulfate followed by dialysis with phosphate buffered saline.
- Taglia:1 mL
- Conf:1 mL