PCR plates, 96-well, fully skirted, exo-frame
Catalog # ISTSIST-601-096GCT
Supplier: iST Scientific Ltd
Specifications
- Colore del telaio:grigio
- N. di pozzetti:96
- Profilo:Low
- Bordo:Fully skirted
- Colore del pozzetto:trasparente
- Conf:50
- Volume pozzetti:0,2 ml
Specifications
About this item
Fully skirted 96-well PCR plate with clear PP wells and grey PC frame.
- Free of DNA, DNase, RNase, and PCR inhibitors
- Low binding PP wells
- Alpha numberic labeling
- Manufactured under the most modern clean room conditions
- Automation friendly
Rigid two-component design eliminates warping and distortion during PCR.
Packaging: Packaged 10 plates per sleeve, 50 plates per box.