GLS 80® wide mouth connection transfer cap system, DURAN®
Codice catalogo 217-0896
Nome del fornitore: DWK Life Sciences
Specifiche
- Descrizione:Screw cap GLS 80, 4 ports with GL 18 thread, EPDM seal
- Seq:03
- Conf:2
Specifiche
Informazioni su questo articolo
PP and PTFE.
- Flexible modular system
- Very good temperature resistance
- Safe transfer of liquid media within a closed and sterile system (evaporation is reduced)
Screw Cap GLS 80 with four ports (GL thread). Five different hose diameters (3,2; 6,0; 8,0; 10,00 and 12,0 mm) can be connected. Sterile pressure equalisation is possible through use of membrane filter. Unused ports can be provided with a blind cap.