Specifications
- Per provette:1,5 ml
- N. di fori:6
- Descrizione:MagRack™ 6
- Conf:1
Specifications
About this item
MagRack 6 is a magnetic rack for small-scale protein purification and sample enrichment with magnetic beads. Each rack consists of anodized aluminium housing (blue), detachable plastic bar (white) containing a neodymium magnet.
- Provides convenience by making it easy perform small-scale purifications and enrichment of sample volumes up to 1,5 ml
- Process up to six samples in parallel
- Allows for optimal performance during protein purifications or enrichments
The anodized aluminium housing assists with electrical conductivity whilst the neodymium magnet is extraordinarily strong, helping with magnetic reaction times as well as optimizing performance during protein purifications / enrichments.
MagRack 6 is complementary to the following magnetic bead-based products: Protein A Mag Sepharose, Protein A Mag Sepharose Xtra, Protein G Mag Sepharose, Protein G Mag Sepharose Xtra, His Mag Sepharose Ni, NHS Mag Sepharose, and Streptavidin Mag Sepharose.
MagRack 6 and MagRack Maxi cover a broad range of sample volumes from low-microliter to high-milliter purification scales, enhancing the flexibility of magnetic beads separations.