Cut resistant gloves, Aracut® Lat
Nome del fornitore: HONEYWELL SAFETY
Certificati
Informazioni su questo articolo
Knitted gloves made from 100% Kevlar® with knitted cuffs and reinforced palms and fingertips, made from blue creped latex.
- Natural latex coating offers excellent flexibility and a secure grip
- Optimum breathability due to ventilated back
- Creped latex coating ensures good mechanical resistance to perforation and abrasion
- Additional coating between the thumb and forefinger reinforces the wear areas and increases the lifespan of the glove
Ideal for handling sharp parts in a dry or damp environment.
Gauge: 10
Colour: blue/beige
EN 388 (3344), Cat. II
Certifications: EN 388:2016 (3X44C), EN 407 (X2XXXX); Cat. II
Specifiche
- Catalog No:
- 112-1610
- 112-1609
- 112-1612
- 112-1611
- Modello:
- Lat
- Lat
- Lat
- Lat
- Ambidestro/mano specifico:
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- Monouso/riutilizzabile:
- riutilizzabile
- riutilizzabile
- riutilizzabile
- riutilizzabile
- Materiale rivestimento interno:
- Kevlar®
- Kevlar®
- Kevlar®
- Kevlar®
- Finitura:
- Coated
- Coated
- Coated
- Coated
- Calibro:
- 10
- 10
- 10
- 10
- Tipo polsino:
- Maglia elastica
- Maglia elastica
- Maglia elastica
- Maglia elastica
- Norma EN:
- EN 388:2016 (3X44C), EN 407 (X2XXXX)
- EN 388:2016 (3X44C), EN 407 (X2XXXX)
- EN 388:2016 (3X44C), EN 407 (X2XXXX)
- EN 388:2016 (3X44C), EN 407 (X2XXXX)