Orders LinkContactUsLinkComponent
SEC standards, AdvanceBio
Nome del fornitore:   AGILENT HEWLETT PACKARD
SEC standards, AdvanceBio
Nome del fornitore:  AGILENT HEWLETT PACKARD
Certificates
SEC standards, AdvanceBio
SEC standards, AdvanceBio
2 varianti
Product Details & Documents
AdvanceBio SEC Protein Standards are uniquely designed for AdvanceBio SEC columns. The standards are manufactured from a select group of proteins and peptides optimized to deliver accurate, precise quantitation for mAb aggregation and protein analysis.
  • Wide range of molecular weight proteins
  • Optimize for use with AdvanceBio SEC columns for maximum peak to peak resolution
  • Manufactured and shipped in moisture and light resistant packaging to ensure stability of standard
This new SEC technology is designed and engineered by Agilent to improve lab productivity by providing robust, reliable methods that eliminate sample reanalysis. Consistent results are achieved from column to column, batch to batch, and lab to lab, ensuring that methods can be transferred across departments and locations to put an end to uncertainty.
Product Family Options
Informazioni sul prodotto
  • Conf
    Disponibilità
    Price
  • HEWL5190-9028
    RNA resolution standard, oligos at 14, 17, 20, and 21 mer
    RNA resolution standard, oligos at 14, 17, 20, and 21 mer
    1
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HEWL5190-9029
    DNA ladder standard, with 15, 20, 25, 30, 35, and 40-mer oligos
    DNA ladder standard, with 15, 20, 25, 30, 35, and 40-mer oligos