SPME fibers
SPME fiber, DVB/PDMS, manual, 65 µm, Stableflex
Codice catalogo VARISU57326U
Nome del fornitore: VARIAN
Vai a
Product Details & Documents
SPME fibers are available in a range of chemistries, formats, and for use with autosamplers or manual injections.
- SPME fibers can be used multiple times when treated with the proper care and caution
- Each fiber has a colour-coded or notched hub, indicating the type of coating on the fiber
Kits are also available to support method development.
Ordering information: When ordering SPME fibers, note that the fiber kits contain fibers only. For a first-time order, you will also need to order the appropriate fiber holder for your needs.
Specifiche
- Descrizione:SPME fiber, DVB/PDMS, manual, 65 µm, Stableflex