Distilling flask with side tube, Assistent
Catalog # HECH41037016
Supplier: GLASWARENFABRIK KARL HECHT
Specifications
- Capacità:250 ml
- Conf:1
Specifications
About this item
This distilling flask is made of borosilicate glass 3.3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion
The length of the side-tube is approximately 200 mm.