Multiple affinity removal spin cartridge HSA/IgG
Multiple affinity removal system spin cartridge, human albumin/IgG, 0,45 ml
Codice catalogo AGLS5188-8825
Nome del fornitore: AGILENT
Multiple affinity removal spin cartridge HSA/IgG
Codice catalogo AGLS5188-8825
Nome del fornitore: AGILENT
Vai a
Product Details & Documents
The multiple affinity removal spin cartridge – Human Albumin/IgG is designed to remove two interfering high-abundant proteins from human samples (such as plasma, serum, urine or cerebrospinal fluid).
- Compatible with standard benchtop microcentrifuges and luer-lok syringes
- Serum load volume is 50 µl per run
- Optimise sample recovery by utilising our rapid, single injection protocols and optimised buffers
Removal of these abundant proteins improves the subsequent LC/MS and electrophoretic analysis of the sample by effectively expanding the dynamic range of the analysis.
Attenzione: For research use only. Not for use in diagnostic procedures.
Specifiche
- Capacità:0,45 ml
- Descrizione:Multiple affinity removal system spin cartridge, human albumin/IgG